Learn More
Medchemexpress LLC (Rac)-Tivantinib | 1239986-50-5 | 95.1% | C23H19N3O2 | 10 MM 1 ML
SDP
Supplier:  Medchemexpress LLC HY7668810MM/1ML
Encompass_Preferred
(Rac)-Tivantinib is a chemical compound closely related to Tivantinib, primarily used as an experimental control in research settings.
  • Isomer of tivantinib
  • Can be utilized as an experimental control
  • Related tivantinib is a highly selective c-Met tyrosine kinase inhibitor
  • Related tivantinib exhibits a Ki of 355 nM
Catalog No. 50-004-34058