Learn More
Medchemexpress LLC (Rac)-Tivantinib | 1239986-50-5 | 95.2% | C23H19N3O2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY76688100MG
Encompass_Preferred
(Rac)-Tivantinib is an isomer of Tivantinib and can be used as an experimental control. Tivantinib is a highly selective c-Met tyrosine kinase inhibitor with a Ki of 355 nM.
  • Isomer of tivantinib
  • Can be used as an experimental control
  • Highly selective c-Met tyrosine kinase inhibitor
  • Ki of 355 nM
Catalog No. 50-004-33994