Learn More
Medchemexpress LLC (Rac)-Tivantinib | 1239986-50-5 | 95.2% | C23H19N3O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY766885MG
Encompass_Preferred
(Rac)-Tivantinib is an isomer of Tivantinib and serves as an experimental control. Tivantinib is identified as a highly selective c-Met tyrosine kinase inhibitor, demonstrating a Ki of 355 nM. This product is intended for research use only.
  • Isomer of Tivantinib.
  • Functions as an experimental control.
  • Related to Tivantinib, a selective c-Met tyrosine kinase inhibitor with a Ki of 355 nM.
  • Available in solid form, off-white to pink color.
Catalog No. 50-004-34175