Learn More
Abcam rac-Tocol, Internal standard for tocopherols and tocotrienols, 1ML
Supplier:  Abcam AB1440671ML
Encompass_Preferred
MW 388.6 Da, Purity >95%. Internal standard for tocopherols and tocotrienols. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6039