Learn More
Medchemexpress LLC (Rac)-TTA-P2 | 918430-49-6 | 98.7% | C21H29Cl2FN2O2 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY50707
Encompass_Preferred
(Rac)-TTA-P2 is an isomer of TTA-P2 and can be used as an experimental control. TTA-P2 is a potent inhibitor of the T-Type calcium channel. It penetrates the Central Nervous System (CNS) effectively and blocks native T-type currents in deep cerebellar nuclear neurons, completely abolishing the window current for both wild-type and mutant Cav3.1 channels. This product has potential for research into neurological diseases.
  • Can be used as an experimental control
  • Potent inhibitor of the T-type calcium channel
  • Effectively penetrates the central nervous system (CNS)
  • Blocks native T-type currents in deep cerebellar nuclear neurons
  • Completely abolishes window current for both wild-type and mutant Cav3.1 channels
  • Has potential for research in neurological diseases
Catalog No. 50-003-57933