Learn More
Medchemexpress LLC (Rac)-TTA-P2 | 918430-49-6 | 98.7% | C21H29Cl2FN2O2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY5070750MG
Encompass_Preferred
(Rac)-TTA-P2 is an isomer of TTA-P2, a potent inhibitor of T-Type calcium channels. It serves as an experimental control and effectively penetrates the central nervous system (CNS). This compound can block native T-type currents in deep cerebellar nuclear neurons, completely abolishing the window current for both wild-type and mutant Cav3.1 channels.
  • Potent inhibitor of T-Type calcium channels.
  • Effectively penetrates the CNS.
  • Blocks native T-type currents in deep cerebellar nuclear neurons.
  • Abolishes window current for wild-type and mutant Cav3.1 channels.
  • Potential for research in neurological diseases.
  • Used as an experimental control.
Catalog No. 50-003-57987