Learn More
Medchemexpress LLC (rac)-viloxazine-d5 hydrochloride | 1276483-10-3 | 278.79 | C13H15D5ClNO3 | 1MG
SDP
Supplier:  Medchemexpress LLC HY125784S1MG
Encompass_Preferred
(rac)-Viloxazine-d5 hydrochloride is the deuterium-labeled racemic form of viloxazine hydrochloride supplied as a stable-isotope reference standard for analytical and research use. It is provided as the hydrochloride salt and intended for use as an internal standard or tracer in mass spectrometry assays; not for human or clinical use. Molecular formula C13H15D5ClNO3, molecular weight 278.79, CAS 1276483-10-3.
  • Deuterium-labeled tracer (d5).
  • Racemic form.
  • Hydrochloride salt.
  • Suitable for liquid chromatography-mass spectrometry internal standard use.
  • Intended for research use only, not for human use.
  • Supplied in a small analytical quantity (1 mg) for tracer and calibration workflows.
Catalog No. 50-002-35139