Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rac-WAY-161503 hydrochloride | 276695-22-8 | MFCD06411596 | 99.5% | 308.59 g/mol | C11H12Cl3N3O | 5 MG
SDP

Supplier:  Medchemexpress LLC HY1031385MG

Encompass_Preferred

(Rac)-WAY-161503 hydrochloride is a racemic research compound that acts as a potent and selective 5-HT2C receptor agonist for neuroscience studies. Supplied as a high-purity solid, it is intended for use in in vitro and in vivo laboratory research.

  • Potent, selective 5-HT2C receptor agonist (Ki = 4 nM; EC50 = 12 nM).
  • High purity (99.5%).
  • Molecular weight 308.59 g/mol.
  • White to off-white solid physical form.
  • CAS registry number 276695-22-8.
  • Store powder at -20°C for long-term stability.
  • Provided in small milligram quantities suitable for research (5 mg typical).

Catalog No. 50-002-46356


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.