Learn More
Medchemexpress LLC (Rac)-X77 | 2144491-78-9 | 99.7% | 459.58 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY136298
Encompass_Preferred
(Rac)-X77 is a racemate of X77, a potent non-covalent inhibitor of the main protease of SARS-CoV-2 (SARS-CoV-2 Mpro). It binds to SARS-CoV-2 Mpro with a Kd value of 0.057 μM, making it a significant compound for research into coronaviruses. This product is a white to off-white solid with a high purity of 99.7%.
  • Potent non-covalent inhibitor of SARS-CoV-2 Mpro
  • Binds to SARS-CoV-2 Mpro with a Kd value of 0.057 μM
  • Identified for laboratory chemical use
  • For research use only
  • Appearance: White to off-white solid
Catalog No. 50-003-52312