Learn More
Medchemexpress LLC Rac-XL177A | 2417089-73-5 | 861.47 | C48H57ClN8O5 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY138794A5MG
Encompass_Preferred
(Rac)-XL177A is the racemic isomer of XL177A, a highly potent and selective irreversible inhibitor of the deubiquitinase USP7. It exhibits sub-nanomolar biochemical potency (reported IC50 ~0.34 nM) and elicits cancer cell-killing effects predominantly through a p53-dependent mechanism, making it suitable for biochemical and cell-based research into USP7 and p53 pathways.
  • Racemic small molecule USP7 inhibitor with sub-nanomolar potency.
  • Irreversible mechanism of action for sustained target engagement.
  • Induces p53-dependent cancer cell killing in cellular assays.
  • Suitable for biochemical and cell-based studies probing deubiquitinase function.
  • Available in multiple vial sizes for screening and follow-up studies.
Catalog No. 50-002-22528