Learn More
Medchemexpress LLC (Rac)-XL177A | 2417089-73-5 | 99.6% | 861.47 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY138794A10MM/1ML
Encompass_Preferred
(Rac)-XL177A is the racemic isomer of XL177A, a highly potent and selective irreversible USP7 inhibitor with an IC50 of 0.34 nM. This compound suppresses cancer cell growth predominantly through a p53-dependent mechanism.
  • Potent and selective irreversible USP7 inhibitor
  • Exhibits an IC50 of 0.34 nM
  • Suppresses cancer cell growth
  • Acts through a p53-dependent mechanism
  • Racemic isomer
Catalog No. 50-003-50496