Learn More
Medchemexpress LLC Rac-zearalenone-d6 | 1185236-04-7 | MFCD28899396 | 324.40 | C18H16D6O5 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY103447S1MG
Encompass_Preferred
(Rac)-Zearalenone-d6 is a deuterium-labeled analogue of the mycotoxin zearalenone supplied as a white to off-white solid for use as a tracer or internal standard in quantitative analytical methods.
  • Deuterium-labeled compound for isotope-dilution analyses.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS quantitation.
  • Provided as a solid with molecular formula C18H16D6O5 and molecular weight 324.40.
  • SDS and certificate of analysis available to confirm specifications.
  • Intended for analytical and reference use; store under conditions specified in the certificate of analysis.
Catalog No. 50-002-45135