Learn More
Medchemexpress LLC (Rac)-zevaquenabant ((Rac)-MRI-1867) | 1610420-28-4 | 99.2% | C25H21ClF3N5O2S | 50 MG

Supplier: Medchemexpress LLC HY14141150MG
(Rac)-Zevaquenabant ((Rac)-MRI-1867) is a cannabinoid receptor type 1 (CB1R)/iNOS antagonist with a Ki of 5.7 nM for CB1R. It has potential for the research of liver fibrosis. It is for research use only and not sold to patients. This compound appears as a white to off-white solid.
- Cannabinoid receptor type 1 (CB1R)/iNOS antagonist
- Ki of 5.7 nM for CB1R
- Potential for the research of liver fibrosis
- For research use only
- Molecular weight of 547.98
- Appears as a white to off-white solid
- Can be shipped at room temperature within the continental US
- Powder storage: -20°C for 3 years; 4°C for 2 years
- In solvent storage: -80°C for 6 months; -20°C for 1 month
- Solubility in DMSO is 50 mg/mL (91.24 mM)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.