Learn More
Medchemexpress LLC Rac-zevaquenabant (Rac-MRI-1867, compound 6b) | 1610420-28-4 | 99.2% | C25H21ClF3N5O2S | 10MG

Supplier: Medchemexpress LLC HY14141110MG
(Rac)-Zevaquenabant is a high-purity research compound that acts as a cannabinoid receptor type 1 (CB1R) and inducible nitric oxide synthase (iNOS) antagonist, used for preclinical pharmacology and disease-model studies including liver fibrosis research. The compound is supplied as a powder with documented solubility and storage recommendations.
- Dual CB1R and iNOS antagonist activity.
- High reported CB1R affinity (Ki 5.7 nM).
- High purity (99.2%).
- Molecular weight 547.98 g/mol.
- Soluble in DMSO; sonication recommended for stock solutions.
- Stable when stored as powder at -20°C for long-term use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.