Learn More
Medchemexpress LLC (Rac)-zlc-002 | 00-00-0 | 99.0% | 231.25 g·mol⁻¹ | C10H17NO5 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1249965MG
Encompass_Preferred
(Rac)-ZLc-002 is a research-grade small molecule inhibitor of the interaction between neuronal nitric oxide synthase (nNOS) and NOS1AP. It has been reported to suppress inflammatory nociception and chemotherapy-induced neuropathic pain, and to act synergistically with paclitaxel to reduce tumor cell viability. The compound is supplied as an off-white to light-yellow solid with high purity and DMSO solubility for in vitro use.
  • Inhibits nNOS-NOS1AP protein-protein interaction
  • Suppresses inflammatory and chemotherapy-induced neuropathic pain
  • Enhances paclitaxel-mediated reduction of tumor cell viability
  • High purity suitable for biological assays
  • Soluble in DMSO for in vitro formulation
  • Stable as a powder under recommended cold storage
Catalog No. 50-002-64617