Learn More
Cayman Chemical racDesethyl OxybutynInhydr 5mg
SDP
Supplier:  Cayman Chemical 342625 MG
Encompass_Preferred
An active metabolite of oxybutynin; binds to mAChRs in isolated rat bladder, submaxillary gland, heart, and colon (Kis = 5.38, 3.12, 16, and 5.59 nM, respectively); reduces micturition pressure in rats at 0.4 and 0.8 mg/kg
Catalog No. 50-294-8633