Learn More
Selleck Chemical LLC Racecadotril S2503-50mg
SDP
Supplier:  Selleck Chemical LLC S250350MG
Encompass_Preferred
Racecadotril (Acetorphan) is a peripherally acting enkephalinase inhibitor with an IC50 of 4 5 M
Catalog No. 50-311-1752