Learn More
Cayman Chemical RacecadotrIl 250mg
SDP
Supplier:  Cayman Chemical 20390250 MG
Encompass_Preferred
A prodrug form of thiorphan; reduces or prevents castor oil-induced diarrhea without delaying intestinal transit in rats at 80 or 100 mg/kg, respectively; decreases duration of diarrhea and increases body weight gain in a neonatal gnotobiotic pig model of human rotavirus-induced diarrhea
Catalog No. 50-298-5543