Learn More
Medchemexpress LLC Racecadotril | 81110-73-8 | 99.9% | 385.48 | 1 G
SDP
Supplier:  Medchemexpress LLC HY173991G
Encompass_Preferred
Racecadotril (Acetorphan) is a neutral endopeptidase (NEP) inhibitor. Its active metabolite Thiorphan inhibits purified NEP activity from mouse brain. This compound acts as an antidiarrheal agent.
  • Neutral endopeptidase inhibitor
  • Inhibits purified NEP activity
  • Acts as an antidiarrheal agent
  • Suitable for various research applications, including cellular and in vivo studies
Catalog No. 50-003-83963