Learn More
Medchemexpress LLC Racecadotril (acetorphan) | 81110-73-8 | 99.94% | 385.48 | 200 MG
SDP
Supplier:  Medchemexpress LLC HY17399200MG
Encompass_Preferred
Racecadotril (Acetorphan) is a neutral endopeptidase (NEP) inhibitor. Racecadotril and its active metabolite Thiorphan inhibit purified NEP activity from mouse brain. It acts as an antidiarrheal agent and is for research use only.
  • Inhibits neutral endopeptidase (NEP) activity
  • Effective in animal models as an antidiarrheal agent
  • High purity of 99.94%
  • Stable for extended periods in powder form
Catalog No. 50-003-84599