Learn More
Medchemexpress LLC Racemate of BL-918 | 2435589-07-2 | 98.9% | 533.44 | C23H15F8N3OS | 50MG
SDP
Supplier:  Medchemexpress LLC HY124729A50MG
Encompass_Preferred
Rac-BL-918 is the racemic form of BL-918, a small-molecule activator of UNC-51-like kinase 1 (ULK1) that induces cytoprotective autophagy. It is supplied for research use in solid and solution formats and is used in cellular assays investigating autophagy and neurodegenerative disease mechanisms. The product is provided with high reported purity suitable for biochemical and cell-based studies.
  • Racemic small-molecule activator of UNC-51-like kinase 1 (ULK1).
  • Induces cytoprotective autophagy in cellular models.
  • Available as solid powder and as DMSO solutions for assay preparation.
  • High reported purity suitable for research (≈98.9%).
  • Used in studies of autophagy and neurodegenerative disease mechanisms.
Catalog No. 50-002-36238