Learn More
Strem, An Ascensus Company CAS# 255836-67-0. 250mg. racemic-2-Di-t-butylphosphino-1,1'-binaphthyl, 98% TrixiePhos. MFCD03094576
SDP
Supplier:  Strem, An Ascensus Company 151043250MG
Encompass
CAS# 255836-67-0. 250mg. racemic-2-Di-t-butylphosphino-1,1'-binaphthyl, 98% TrixiePhos. MFCD03094576. Molecular Weight: 398.53. Molecular Formula: C28H31P. Color/form: white xtl. Strem# 15-1043. http://www.strem.com/catalog/v/15-1043/
Catalog No. 50-901-10034