Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Racemic anabasine | 13078-04-1 | 162.24 g/mol | C10H14N2 | 100 MG
SDP

Supplier:  Medchemexpress LLC HYW052144100MG

Encompass_Preferred

(±) Anabasine is the racemic form of anabasine, an alkaloid that acts as an agonist of the α7 nicotinic acetylcholine receptor and has reported anti-inflammatory and insecticidal activities. See manufacturer datasheet for solubility, storage, and handling details.

  • Racemic form of anabasine
  • Agonist of α7 nicotinic acetylcholine receptor
  • Exhibits anti-inflammatory and insecticidal activities
  • CAS number 13078-04-1
  • Molecular formula C10H14N2; molecular weight 162.24 g/mol
  • Solubility: in vitro DMSO 130 mg/mL; in vivo formulations ≥ 2.17 mg/mL (clear solutions)
  • Storage: pure form -20°C (3 years); 4°C (2 years)

Catalog No. 50-002-10014


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.