Learn More
Medchemexpress LLC Racemic bepotastine besilate | 1415692-17-9 | 99.5% | 547.06 | C27H31ClN2O6S | 10 MG

Supplier: Medchemexpress LLC HYA0015A10MG
Racemic bepotastine besilate is a high-purity research compound used as an experimental control in studies of histamine H1 receptor activity and allergic disease mechanisms. It is provided with supporting analytical data for laboratory research use only.
- High purity (99.52%) suitable for research applications.
- Molecular weight 547.06 g/mol.
- Molecular formula C27H31ClN2O6S.
- CAS number 1415692-17-9.
- Supplied as a 10 mg package for laboratory use.
- Intended for research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.