Learn More
Medchemexpress LLC Racemic pyrotinib | 1246089-97-3 | 99.3% | 583.08 | C32H31ClN6O3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY104065A10MG
Encompass_Preferred
Racemic pyrotinib is the racemate of pyrotinib, a potent and selective dual EGFR/HER2 receptor tyrosine kinase inhibitor intended for research use. It is supplied as a powder for in vitro studies and preclinical assays, with documented solubility and storage recommendations.
Catalog No. 50-002-43517