Learn More
Medchemexpress LLC Racemic voriconazole | 188416-29-7 | MFCD09263753 | 99.1% | 349.31 g/mol | C16H14F3N5O | 25 MG

Supplier: Medchemexpress LLC HYW33756925MG
(±)-Voriconazole (racemic voriconazole) is the racemic form of the second-generation triazole antifungal, supplied for research use. It acts as a broad-spectrum inhibitor of fungal ergosterol biosynthesis and is provided as a high-purity research chemical.
- Racemic configuration (mixture of enantiomers).
- Second-generation, broad-spectrum antifungal activity.
- Inhibits fungal ergosterol biosynthesis.
- High purity (99.06%).
- Molecular weight 349.31 g/mol; formula C16H14F3N5O.
- Provided as a research-grade chemical for laboratory use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.