Learn More
Medchemexpress LLC Racemic voriconazole | 188416-29-7 | MFCD09263753 | 99.1% | 349.31 g/mol | C16H14F3N5O | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW33756950MG
Encompass_Preferred
(±)-Voriconazole is the racemic form of voriconazole, a triazole antifungal agent supplied as a white to off-white solid for laboratory research. It has molecular formula C16H14F3N5O, a molecular weight of 349.31 g/mol, and CAS number 188416-29-7. The material is provided in small milligram packaging for analytical and experimental use.
  • High purity (99.06%) for reliable experimental results.
  • White to off-white solid, easy to weigh and handle.
  • Well-characterized chemical identifiers for accurate substance identification.
  • Suitable for antifungal mechanism and biochemical studies.
  • Provided in small, controlled quantities for research workflows.
Catalog No. 50-002-81851