Learn More
Medchemexpress LLC Racemic WAY-161503 | 75704-24-4 | MFCD06411596 | 98.5% | 272.13 g/mol | C11H11Cl2N3O | 1 ML
SDP
Supplier:  Medchemexpress LLC HY103138A10MM 1ML
Encompass_Preferred
(Rac)-WAY-161503 is a racemic 5-HT2C receptor agonist supplied for laboratory research to study serotonergic signaling, feeding behavior, and mood-related pathways. The compound is offered as a solid (powder) and as a ready-to-use 10 mM solution in DMSO (1 mL). Chemical identifiers include CAS 75704-24-4, molecular formula C11H11Cl2N3O, and molecular weight 272.13 g/mol. For research use only.
  • Potent 5-HT2C receptor agonist (Ki ≈ 4 nM; EC50 ≈ 12 nM).
  • Available as powder and as a 10 mM solution in DMSO, 1 mL.
  • High purity (≈98.5% by HPLC).
  • Soluble in DMSO (100 mg/mL); instructions provided for preparing 1, 5, and 10 mM stocks.
  • Suitable for in vitro pharmacology and receptor signaling studies.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-47807