Learn More
U.S. Pharmacopeia Racepinephrine Hydrochloride, 329-63-5, MFCD00050562, 30mg
Supplier:  U.S. Pharmacopeia 1598097
Encompass
Molecular formula C9H13NO3.ClH, Molecular weight 219.67, Melting point/freezing point 311 - 314.6 °F (155 - 157 °C), Synonyms: DL-Adrenaline hydrochloride
Catalog No. NC1710956