Learn More
Thomas Scientific 81 Place (9x9), white, for up to 2" tall x 13mm wide tubes
SDP
Supplier:  Thomas Scientific 1216M64
Encompass
Globe Scientific's cardboard storage boxes are designed for the compact storage of sample tubes, cryogenic vials and centrifuge tubes in a temperature range of -196°to +121°C. They have a liquid resistant coating and are designed for use in refrigerator and cryogenic freezer systems.
Catalog No. NC0663108