Learn More
Cole-Parmer Upright Freezer Drawer Rack for 96 Deep-Well Plates or 1 3/4" Max H Boxes, 7 x 5 Array
Supplier:  Cole-Parmer 0440084
Encompass_Preferred
Drawers provide quick storage and retrieval of plates and microtube boxes. Easily access fewer microtiter plates and boxes at a time, protecting other samples in the rack from warming up. Durable corrosion-resistant stainless steel construction for long-term use.
Catalog No. NC1568907