Learn More
Chemglass Life Sciences Chemglass 50-Place Low-Profile Tube Rack with Grippers, Blue, for up to 17 mm Tubes
SDP
Supplier:  Chemglass Life Sciences CLS477915
Encompass_Preferred
The Chemglass 50-place low-profile rack secures tubes with integrated rubber grippers and is molded from reinforced, autoclavable nylon for chemical and thermal resistance. It is designed for general tube storage, handling, and decanting, and features a flat plate area for labeling.
  • Low-profile design for easy label visibility.
  • Holds up to 50 tubes in a 5 x 10 layout.
  • Rubber grippers hold tubes during decanting.
  • Molded reinforced autoclavable nylon construction.
  • Compatible with tubes up to 17 mm diameter.
  • Flat plate area for labeling.
  • Will not float in a water bath.
Catalog No. NC2943826