Learn More
Chemglass Life Sciences Chemglass Rack with Grippers, Low Profile, 50-Place (for up to 17 mm tubes) - Orange
SDP
Supplier:  Chemglass Life Sciences CLS477917
Encompass_Preferred
This low-profile tube rack holds and secures up to 50 tubes with rubber grippers designed to prevent slippage during decanting and handling. The rack is autoclavable and chemically resistant, making it suitable for routine laboratory storage and organization.
  • Low-profile design for easy label viewing.
  • Holds 50 tubes in a 5 x 10 configuration.
  • Holds tubes up to 17 mm diameter.
  • Rubber grippers secure tubes during decanting.
  • Reinforced autoclavable nylon construction with POM grippers.
  • Will not float in a water bath.
  • Flat plate area on the end for labeling.
  • Excellent thermal and chemical resistance.
Catalog No. NC2943827