Learn More
Uline Rackable Plastic Pallet 48x40"""" Blue
Supplier:  Uline H8933
Encompass
48 x 40"""" blue plastic pallet, weight capacity: floor 30k lbs, fork 5k lbs, rack 2,200 lbs, HDPE
Catalog No. NC1971127