Learn More
Apexbio Technology LLC Raclopride 84225-95-6 50mg
SDP
Supplier:  Apexbio Technology LLC B686450
Encompass_Preferred
Raclopride is a small-molecule antagonist targeting dopamine D2 and D3 receptors It is designed to competitively bind these receptors thereby inhibiting dopaminergic neurotransmission and influencing downstream signaling pathways Raclopride exerts its biological activity primarily through competitive antagonism of the dopamine D2 (Ki 1 8 nM) and D3 receptors (Ki 3 5 nM) Based on these pharmacological properties Raclopride holds research potential in neuroscientific studies investigating dopamine-related neurological conditions such as the underlying molecular mechanisms of schizophrenia and associated psychiatric disorders and in exploring receptor-specific functions in dopaminergic signaling
Catalog No. 50-000-89282