Learn More
Strem, An Ascensus Company CAS# [121009-93-6]. 100mg. rac-Dimethylsilylbis(1-indenyl)zirconium dichloride, min. 97%. MFCD00672108
SDP
Supplier:  Strem, An Ascensus Company 401142100MG
Encompass
CAS# [121009-93-6]. 100mg. rac-Dimethylsilylbis(1-indenyl)zirconium dichloride, min. 97%. MFCD00672108. Molecular Weight: 448.53. Molecular Formula: (CH3)2Si(C9H6)2ZrCl2. Color/form: orange pwdr. Strem# 40-1142. www.strem.com/catalog/v3/40-1142/
Catalog No. 50-197-9378