Learn More
Cayman Chemical RactopamInd3hydrochLRIde 10mg
SDP
Supplier:  Cayman Chemical 3720710 MG
Encompass_Preferred
An internal standard for the quantification of ractopamine by GC- or LC-MS
Catalog No. 50-291-5096