Learn More
Cayman Chemical RAD17dRIved PeptIDtrIflu 10mg
SDP
Supplier:  Cayman Chemical 3751210 MG
Encompass_Preferred
A peptide substrate for ATR; has been used to identify inhibitors of ATR
Catalog No. 50-291-5692