Learn More
Santa Cruz Biotechnology Rad51 Antibody (3C10), 200 µg/ml
Supplier:  Santa Cruz Biotechnology SC53428
Encompass
Rad51 Antibody (3C10) is an IgG1 λ mouse monoclonal Rad51 antibody that detects Rad51 of mouse, rat and human origin.
Catalog No. NC2754942