Learn More
Aviva Systems Biology Corp RAD51L3 Antibody - C-terminal region (ARP61547_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61547_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RAD51L3. It was validated on Western Blot. . Alternate names: HsTRAD, R51H3, RAD51D, Trad, TRAD, BROVCA4, RAD51L3ApplicationWB. Species reactivity: Human, Rat
Catalog No. 50-287-406