Learn More
Selleck Chemical LLC Raddeanin A S9262-5MG
SDP
Supplier:  Selleck Chemical LLC S92625MG
Encompass_Preferred
Raddeanin A (Raddeanin R3 NSC382873) a triterpenoid saponin from Anemone raddeana Regel displays moderate inhibitory activity against histone deacetylases (HDACs) and has high antiangiogenic potency antitumor activity
Catalog No. 50-000-23723