Learn More
Cayman Chemical Raddenn A 50mg
SDP
Supplier:  Cayman Chemical 4023550 MG
Encompass_Preferred
A triterpenoid saponin with diverse biological activities; binds to Aβ42 (Kd = 27.63 µM); inhibits the invasion and migration of, and induces apoptosis and cell cycle arrest at the G0/G1 phase in, HeLa cervical cancer cells at 4 µM; improves learning and memory in the Morris water maze in mice in the 3xTg model of Alzheimer’s disease at 10 mg/kg; reduces neurological deficits, lesion volume, edema, and microglia lesion infiltration in a mouse model of acute cerebral hemorrhage induced by intracerebral injection of bacterial collagenase VII
Catalog No. 50-294-3812