Learn More
Cayman Chemical Radicicol, 12772-57-5, 1 mg
SDP
Supplier:  Cayman Chemical 130891
Encompass_Preferred
A natural product with antibiotic, antifungal, and antimalarial properties; binds and inhibits heat shock protein 90 (HSP90) with nanomolar affinity
Catalog No. NC0472403