Learn More
Medchemexpress LLC HY-109574 5mg Medchemexpress, Raf inhibitor 2 CAS:220904-99-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1095745MG
Encompass_Preferred
Medchemexpress, HY-109574 5mg Raf inhibitor 2 CAS:220904-99-4 Raf inhibitor 2 is a potent raf kinase (IC50<1.0 μM) inhibitor, compound 32, extracted from patent EP1003721B1. Raf inhibitor 2 can be used for cancer research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0773