Learn More
Selleck Chemical LLC Raf inhibitor 2-E0934-5MG
SDP
Supplier:  Selleck Chemical LLC E09345MG
Encompass_Preferred
Raf inhibitor 2 is a potent Raf kinase inhibitor with an IC50 of 1 0 M
Catalog No. 50-313-8747