Learn More
ECM Bioscience Raf Phospho-Regulation Antibody Sampler Kit, ECM Biosciences
Supplier:  ECM Bioscience RK6080
Encompass_Preferred
The Raf phospho-regulation antibody sampler kit can be used to examine phosphorylation of A-Raf, B-Raf, and C-Raf. The anti-B-Raf (Ser-446) detects site-specific phosphorylation of B-Raf, as well as the conserved site in C-Raf (Ser-338) and A-Raf (Ser-299). Similarly, the anti-C-Raf (Ser-471) detects site-specific phosphorylation in C-Raf, as well as in B-Raf (Ser-579) and A-Raf (Ser-432). The kit also includes antibodies to monitor total expression levels of A-Raf, B-Raf, and C-Raf. Applications: WB, E; Species Reactivity: Hu, Rt, Ms
Catalog No. 50-206-8812