Learn More
TARGETMOL CHEMICALS INC RAGAGLITAZAR 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T285015MG
Encompass_Preferred
Also available in 50 mg 100 mg and bulk. Please contact Fisher for quotes. Ragaglitazar(NNC-61-0029) is a potent dual PPAR(alpha) and PPAR(gamma) agonist that has shown potent lipid-lowering and insulin sensitizing effects in animal models.Ragaglitazar can be used to study type 2 diabetes. purity: 99%
Catalog No. 50-258-0510