Learn More
Pipette Supplies, Inc. Rainin Pipet-Lite O-ring, Single & Multichannel, 300μL (Pipette Supplies)
SDP
Supplier:  Pipette Supplies, Inc. 20300/5
Encompass
Piston O-ring for the following Rainin 300μL Pipettes:Pipet-Lite Single Channel (L, SL)Pipet-Lite Multichannel (L8/L12)Pipet-Plus (R, RL)EDP1 (E1, SE1)EDP3, EDP3 Plus Single Channel (E3, SE3)EDP3, EDP3 Plus Multichannel (E8/E12)EDP3 ASM Adjust Space Multichannel (EA6, EA8)E-Man Hybrid (H, SH)Pipet-Lite XLS Single Channel XLS (XLS)Pipet-Lite XLS Multichannel (L8/L12XLS)E4 XLS Single Channel (E4, SE4)E4 XLS Multichannel (E8/E12XLS)Alternative to Rainin 6200-414 / 17007532
Catalog No. NC1908813