Learn More
ATCC RAJI-LUC2
Supplier:  ATCC CCL86LUC2/DEL
Encompass
Raji-Luc2 CAR-T Target Luciferase Reporter Cells can be used as a target cancer cell for in vitro killing assay by CD19 CAR-T cells (tested at ATCC) and is expected to also work for CD20 CAR-T cells. Raji-Luc2 naturally expresses high levels of CD19 (verified at ATCC).
Catalog No. NC2198000