Learn More
AMID BIOSCIENCES Human RalA Protein, Recombinant, Biotinylated, 0.1 mg
SDP
Supplier:  AMID BIOSCIENCES RALAB301
Encompass_Preferred
Human RalA is supplied as a recombinant biotinylated small GTPase in a 0.1 mg format. RalA is a RAS superfamily protein used in studies of small GTPase signaling effector binding vesicle trafficking and oncogenic. Applications include SPR, BLI, pull-down, AlphaScreen, RAL-family GTPase signaling studies, RAS-effector pathway assays, small molecule screening. Technical specifications are included below.- Quantity - 0.1 mg- Gene - RALA- Mutation / Variant - Wild-type- Organism - Human- Tag - N-terminal 8XHis, N-terminal AviTag- Expression System - E. coli- Purity - greater than 90% by Coomassie staining- UniProt ID - P11233- Amino Acid Range - 1-206- Buffer - 20 mM Tris-HCl, pH 7.5, 100 mM NaCl, 2 mM DTT, 10% Glycerol- Endotoxin Level - less than 1 EU/mg- Storage - -80 C- Biotinylated - Yes, N-terminusThis product is for research use only.
Catalog No. NC4043441